Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203853.2 | NP_001190782.1 | 2 - 3 | 357 | 75 | |
NM_118209.3 | NP_567616.1 | 1 - 2 | 502 | 75 | ||
GenBank mRNA | AF327068.1 | - | 1 - 2 | 284 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |