Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118225.5 | NP_193839.4 | 10 - 11 | 2293 | 81 | |
GenBank mRNA | AF515728.1 | - | 10 - 11 | 2160 | 81 | |
AY081328.1 | - | 10 - 11 | 2206 | 81 | ||
BT010547.1 | - | 10 - 11 | 2110 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |