Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118336.5 | NP_193946.3 | 3 - 4 | 595 | 115 | |
GenBank mRNA | AK176241.1 | - | 3 - 4 | 535 | 115 | |
AY572783.1 | - | 3 - 4 | 526 | 115 | ||
BT010440.1 | - | 3 - 4 | 526 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |