Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036621.3 | NP_001031698.1 | 6 - 7 | 1222 | 100 | |
NM_001203871.1 | NP_001190800.1 | 6 - 7 | 1233 | 100 | ||
NM_001341552.1 | NP_001328217.1 | 6 - 7 | 1836 | 100 | ||
NM_001341553.1 | NP_001328218.1 | 6 - 7 | 1647 | 100 | ||
NM_118380.3 | NP_567662.1 | 6 - 7 | 1870 | 100 | ||
NM_202863.3 | NP_974592.1 | 6 - 7 | 1236 | 100 | ||
GenBank mRNA | AK229811.1 | - | 6 - 7 | 1200 | 100 | |
AY052361.1 | - | 6 - 7 | 1755 | 100 | ||
BT001093.1 | - | 6 - 7 | 1503 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |