Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036621.3 | NP_001031698.1 | 7 - 8 | 1370 | 94 | |
NM_001203871.1 | NP_001190800.1 | 7 - 8 | 1381 | 94 | ||
NM_001341552.1 | NP_001328217.1 | 7 - 8 | 1984 | 94 | ||
NM_001341553.1 | NP_001328218.1 | 7 - 8 | 1795 | 94 | ||
NM_118380.3 | NP_567662.1 | 7 - 8 | 2018 | 94 | ||
NM_202863.3 | NP_974592.1 | 7 - 8 | 1384 | 94 | ||
GenBank mRNA | AK229811.1 | - | 7 - 8 | 1348 | 94 | |
AY052361.1 | - | 7 - 8 | 1903 | 94 | ||
BT001093.1 | - | 7 - 8 | 1651 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |