Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT4G22755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118403.2 | NP_567669.1 | 3 - 4 | 921 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |