Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341566.1 | NP_001327985.1 | 7 - 8 | 958 | 74 | |
NM_118422.4 | NP_194024.1 | 7 - 8 | 958 | 74 | ||
GenBank mRNA | AF000146.1 | - | 7 - 8 | 868 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |