Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035888.2 | NP_001030965.1 | 4 - 5 | 702 | 88 | |
NM_126420.2 | NP_565304.1 | 5 - 6 | 569 | 88 | ||
GenBank mRNA | AK317500.1 | - | 4 - 5 | 684 | 88 | |
BT000908.1 | - | 3 - 4 | 307 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |