Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341092.1 | NP_001319956.1 | 11 - 12 | 1535 | 66 | |
NM_117725.3 | NP_193363.4 | 11 - 12 | 1122 | 66 | ||
NM_148347.2 | NP_680713.2 | 12 - 13 | 1331 | 66 | ||
NM_179211.3 | NP_849542.2 | 10 - 11 | 3766 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |