Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341313.1 | NP_001319991.1 | 4 - 5 | 409 | 136 | |
NM_001341314.1 | NP_001319990.1 | 4 - 5 | 622 | 136 | ||
NM_001341315.1 | NP_001329781.1 | 4 - 5 | 409 | 136 | ||
NM_001341316.1 | NP_001329780.1 | 4 - 5 | 528 | 136 | ||
NM_148354.4 | NP_680720.1 | 4 - 5 | 364 | 136 | ||
NM_202841.2 | NP_974570.1 | 4 - 5 | 528 | 136 | ||
GenBank mRNA | AK317105.1 | - | 4 - 5 | 511 | 136 | |
AY065055.1 | - | 4 - 5 | 364 | 136 | ||
AY093752.1 | - | 3 - 4 | 253 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |