Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036038.2 | NP_001031115.1 | 8 - 9 | 1370 | 75 | |
NM_001332891.1 | NP_001320924.1 | 7 - 8 | 1606 | 75 | ||
NM_102771.4 | NP_174323.1 | 7 - 8 | 1475 | 75 | ||
GenBank mRNA | AF013467.1 | - | 7 - 8 | 1400 | 75 | |
AY669788.1 | - | 7 - 8 | 654 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |