Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102785.5 | NP_174337.1 | 14 - 15 | 2170 | 66 | |
GenBank mRNA | AJ440967.1 | - | 14 - 15 | 1821 | 66 | |
AK226570.1 | - | 14 - 15 | 2035 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |