Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102887.5 | NP_174433.2 | 5 - 6 | 879 | 62 | |
GenBank mRNA | AB073133.1 | - | 5 - 6 | 729 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |