Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSYP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333003.1 | NP_001320542.1 | 4 - 5 | 748 | 80 | |
NM_102960.2 | NP_174506.1 | 4 - 5 | 748 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |