Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204026.2 | NP_001190955.1 | 2 - 3 | 369 | 72 | |
NM_001342505.1 | NP_001328584.1 | 1 - 2 | 564 | 72 | ||
NM_001342506.1 | NP_001328583.1 | 2 - 3 | 411 | 72 | ||
NM_120018.4 | NP_195569.1 | 2 - 3 | 367 | 72 | ||
GenBank mRNA | AY086801.1 | - | 2 - 3 | 267 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |