Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHA-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120068.5 | NP_568051.1 | 3 - 4 | 435 | 115 | |
GenBank mRNA | AK227321.1 | - | 3 - 4 | 402 | 115 | |
AY060557.1 | - | 3 - 4 | 396 | 115 | ||
BT002615.1 | - | 3 - 4 | 321 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |