Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342527.1 | NP_001328715.1 | 9 - 10 | 1059 | 79 | |
NM_120081.2 | NP_195632.1 | 9 - 10 | 1237 | 79 | ||
GenBank mRNA | AY059862.1 | - | 9 - 10 | 1177 | 79 | |
BT010378.1 | - | 8 - 9 | 1018 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |