Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204036.1 | NP_001190965.1 | 1 - 2 | 526 | 107 | |
NM_001342538.1 | NP_001320167.1 | 1 - 2 | 423 | 107 | ||
NM_120097.3 | NP_568058.1 | 1 - 2 | 457 | 107 | ||
GenBank mRNA | AF302675.1 | - | 1 - 2 | 287 | 107 | |
AF370208.1 | - | 1 - 2 | 422 | 107 | ||
AY133770.1 | - | 1 - 2 | 269 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |