Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161045.2 | NP_001154517.1 | 7 - 8 | 612 | 133 | |
NM_001202597.1 | NP_001189526.1 | 7 - 8 | 612 | 133 | ||
NM_001335430.1 | NP_001324683.1 | 7 - 8 | 612 | 133 | ||
NM_001335431.1 | NP_001324684.1 | 7 - 8 | 612 | 133 | ||
NM_001335432.1 | NP_001324685.1 | 6 - 7 | 742 | 133 | ||
NM_001335433.1 | NP_001324686.1 | 6 - 7 | 700 | 133 | ||
NM_001335434.1 | NP_001324682.1 | 1 - 2 | 93 | 133 | ||
NM_127032.2 | NP_179075.1 | 6 - 7 | 742 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |