Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161045.2 | NP_001154517.1 | 10 - 11 | 921 | 90 | |
NM_001202597.1 | NP_001189526.1 | 10 - 11 | 921 | 90 | ||
NM_001335430.1 | NP_001324683.1 | 10 - 11 | 921 | 90 | ||
NM_001335431.1 | NP_001324684.1 | 10 - 11 | 921 | 90 | ||
NM_001335432.1 | NP_001324685.1 | 9 - 10 | 1051 | 90 | ||
NM_001335433.1 | NP_001324686.1 | 9 - 10 | 1009 | 90 | ||
NM_001335434.1 | NP_001324682.1 | 4 - 5 | 402 | 90 | ||
NM_127032.2 | NP_179075.1 | 9 - 10 | 1051 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |