Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335508.1 | NP_001318235.1 | 12 - 13 | 1253 | 100 | |
NM_001335509.1 | NP_001323497.1 | 12 - 13 | 1319 | 100 | ||
NM_001335512.1 | NP_001323502.1 | 10 - 11 | 998 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |