Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036289.1 | NP_001031366.1 | 7 - 8 | 1251 | 97 | |
NM_001036290.2 | NP_001031367.1 | 7 - 8 | 1350 | 97 | ||
NM_001335556.1 | NP_001324196.1 | 6 - 7 | 1278 | 97 | ||
NM_001335557.1 | NP_001324198.1 | 6 - 7 | 1387 | 97 | ||
NM_001335558.1 | NP_001324199.1 | 5 - 6 | 1642 | 97 | ||
NM_001335559.1 | NP_001324200.1 | 6 - 7 | 1565 | 97 | ||
NM_001335561.1 | NP_001324195.1 | 6 - 7 | 1528 | 97 | ||
NM_127332.3 | NP_179369.2 | 7 - 8 | 1350 | 97 | ||
GenBank mRNA | AB196961.1 | - | 5 - 6 | 1101 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |