Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337010.1 | NP_001324372.1 | 3 - 4 | 1822 | 72 | |
NM_180051.3 | NP_850382.1 | 4 - 5 | 2122 | 72 | ||
GenBank mRNA | AF474176.1 | - | 4 - 5 | 1939 | 72 | |
BT046199.1 | - | 3 - 4 | 1639 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |