Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337082.1 | NP_001324514.1 | 10 - 11 | 783 | 94 | |
NM_001337083.1 | NP_001324515.1 | 10 - 11 | 691 | 94 | ||
NM_129998.3 | NP_566016.1 | 10 - 11 | 732 | 94 | ||
NM_180084.2 | NP_850415.1 | 10 - 11 | 735 | 94 | ||
GenBank mRNA | AF387018.1 | - | 10 - 11 | 734 | 94 | |
BT006613.1 | - | 9 - 10 | 600 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |