Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035552.2 | NP_001030629.1 | 4 - 5 | 1250 | 63 | |
NM_111266.2 | NP_566218.1 | 4 - 5 | 982 | 63 | ||
NM_180179.3 | NP_850510.1 | 4 - 5 | 1238 | 63 | ||
GenBank mRNA | AF419591.1 | - | 4 - 5 | 981 | 63 | |
AK317103.1 | - | 4 - 5 | 1137 | 63 | ||
AY097342.1 | - | 4 - 5 | 882 | 63 | ||
AY894117.1 | - | 4 - 5 | 1034 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |