Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111458.5 | NP_187235.1 | 10 - 11 | 1520 | 67 | |
NM_180189.4 | NP_850520.1 | 10 - 11 | 1248 | 67 | ||
GenBank mRNA | AK227034.1 | - | 10 - 11 | 1288 | 67 | |
AK316673.1 | - | 10 - 11 | 1317 | 67 | ||
AY062713.1 | - | 10 - 11 | 1233 | 67 | ||
AY093347.1 | - | 9 - 10 | 1042 | 67 | ||
X75431.1 | - | 1215 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |