Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111573.5 | NP_187349.2 | 2 - 3 | 396 | 131 | |
NM_180197.2 | NP_850528.1 | 2 - 3 | 349 | 131 | ||
GenBank mRNA | BT015414.1 | - | 2 - 3 | 258 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |