Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338026.1 | NP_001327857.1 | 5 - 6 | 701 | 101 | |
NM_001338027.1 | NP_001327858.1 | 4 - 5 | 967 | 101 | ||
NM_001338028.1 | NP_001327859.1 | 4 - 5 | 586 | 101 | ||
NM_001338029.1 | NP_001327860.1 | 4 - 5 | 730 | 101 | ||
NM_112143.4 | NP_187912.2 | 4 - 5 | 606 | 101 | ||
NM_180241.2 | NP_850572.1 | 4 - 5 | 586 | 101 | ||
GenBank mRNA | AK228498.1 | - | 4 - 5 | 386 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |