Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338339.1 | NP_001326233.1 | 15 - 16 | 1886 | 82 | |
NM_112737.3 | NP_566612.1 | 17 - 18 | 1741 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |