Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338537.1 | NP_001327775.1 | 2 - 3 | 370 | 72 | |
NM_113045.3 | NP_566686.2 | 2 - 3 | 272 | 72 | ||
NM_180289.4 | NP_850620.2 | 2 - 3 | 370 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |