Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204043.2 | NP_001190972.1 | 12 - 13 | 2676 | 69 | |
NM_001204044.1 | NP_001190973.1 | 12 - 13 | 2673 | 69 | ||
NM_001342558.1 | NP_001328232.1 | 11 - 12 | 3040 | 69 | ||
NM_120148.3 | NP_568072.1 | 12 - 13 | 2663 | 69 | ||
GenBank mRNA | AF378120.1 | - | 12 - 13 | 2592 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |