Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342578.1 | NP_001332650.1 | 3 - 4 | 546 | 113 | |
NM_120197.4 | NP_195739.2 | 3 - 4 | 588 | 113 | ||
GenBank mRNA | BT014855.1 | - | 3 - 4 | 488 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |