Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342587.1 | NP_001330013.1 | 18 - 19 | 3403 | 104 | |
NM_001342588.1 | NP_001330014.1 | 19 - 20 | 3360 | 104 | ||
NM_120218.2 | NP_195760.1 | 19 - 20 | 3145 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |