Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342607.1 | NP_001318456.1 | 7 - 8 | 3498 | 68 | |
NM_001342608.1 | NP_001330632.1 | 5 - 6 | 3434 | 68 | ||
NM_001342609.1 | NP_001330631.1 | 6 - 7 | 3380 | 68 | ||
NM_001342610.1 | NP_001330637.1 | 6 - 7 | 3589 | 68 | ||
NM_001342611.1 | NP_001330636.1 | 5 - 6 | 3434 | 68 | ||
NM_001342612.1 | NP_001330633.1 | 4 - 5 | 2991 | 68 | ||
NM_001342613.1 | NP_001330635.1 | 1 - 2 | 2500 | 68 | ||
NM_001342614.1 | NP_001330634.1 | 6 - 7 | 3589 | 68 | ||
NM_001342615.1 | NP_001330639.1 | 5 - 6 | 3434 | 68 | ||
NM_001342616.1 | NP_001330638.1 | 6 - 7 | 3339 | 68 | ||
NM_120251.4 | NP_195793.3 | 6 - 7 | 3380 | 68 | ||
GenBank mRNA | AY743927.1 | - | 6 - 7 | 3259 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |