Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342696.1 | NP_001330384.1 | 6 - 7 | 823 | 81 | |
NM_001342697.1 | NP_001330385.1 | 5 - 6 | 976 | 81 | ||
NM_120408.4 | NP_195950.1 | 6 - 7 | 605 | 81 | ||
GenBank mRNA | AF180895.1 | - | 6 - 7 | 507 | 81 | |
AY042855.1 | - | 6 - 7 | 585 | 81 | ||
AY072485.1 | - | 6 - 7 | 474 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |