Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCSLA09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120457.4 | NP_195996.1 | 5 - 6 | 1465 | 75 | |
GenBank mRNA | AY060563.1 | - | 5 - 6 | 1385 | 75 | |
AY149935.1 | - | 5 - 6 | 922 | 75 | ||
KJ138709.1 | - | 5 - 6 | 922 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |