Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120476.5 | NP_196014.1 | 8 - 9 | 1086 | 85 | |
GenBank mRNA | AY050999.1 | - | 8 - 9 | 982 | 85 | |
BT000976.1 | - | 8 - 9 | 918 | 85 | ||
Z21970.1 | - | 941 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |