Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127383.3 | NP_179417.1 | 1 - 2 | 310 | 97 | |
GenBank mRNA | AK176897.1 | - | 1 - 2 | 225 | 97 | |
BT010494.1 | - | 1 - 2 | 107 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |