Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDH1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127401.3 | NP_179435.1 | 1 - 2 | 126 | 111 | |
GenBank mRNA | AK119142.1 | - | 1 - 2 | 105 | 111 | |
BT005938.1 | - | 1 - 2 | 102 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |