Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127505.5 | NP_179537.1 | 5 - 6 | 734 | 149 | |
GenBank mRNA | AY086262.1 | - | 5 - 6 | 666 | 149 | |
AY099837.1 | - | 5 - 6 | 625 | 149 | ||
AY943844.1 | - | 5 - 6 | 427 | 149 | ||
BT006580.1 | - | 5 - 6 | 427 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |