Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335672.1 | NP_001325368.1 | 28 - 29 | 3537 | 123 | |
NM_001335673.1 | NP_001325369.1 | 27 - 28 | 3492 | 123 | ||
NM_127588.3 | NP_179619.2 | 28 - 29 | 3346 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |