Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIM44-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335690.1 | NP_001325326.1 | 8 - 9 | 1089 | 149 | |
NM_001335691.1 | NP_001325325.1 | 8 - 9 | 1080 | 149 | ||
NM_001335692.1 | NP_001325324.1 | 8 - 9 | 971 | 149 | ||
NM_127610.1 | NP_565473.1 | 8 - 9 | 971 | 149 | ||
GenBank mRNA | DQ446533.1 | - | 8 - 9 | 977 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |