Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pEARLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335728.1 | NP_001323931.1 | 3 - 4 | 573 | 96 | |
NM_001335729.1 | NP_001323932.1 | 3 - 4 | 573 | 96 | ||
NM_001335730.1 | NP_001323930.1 | 2 - 3 | 747 | 96 | ||
NM_127665.3 | NP_179692.1 | 3 - 4 | 541 | 96 | ||
GenBank mRNA | AY059813.1 | - | 3 - 4 | 535 | 96 | |
BT000010.1 | - | 2 - 3 | 401 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |