Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127685.4 | NP_179711.2 | 4 - 5 | 661 | 70 | |
GenBank mRNA | BT000912.1 | - | 4 - 5 | 418 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |