Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339632.1 | NP_001325690.1 | 3 - 4 | 1072 | 90 | |
NM_115238.2 | NP_566989.1 | 4 - 5 | 1002 | 90 | ||
NM_180367.2 | NP_850698.1 | 3 - 4 | 1072 | 90 | ||
GenBank mRNA | BT029533.1 | - | 2 - 3 | 322 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |