Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125707.2 | NP_001119179.1 | 16 - 17 | 2064 | 97 | |
NM_120743.3 | NP_568171.1 | 16 - 17 | 2237 | 97 | ||
NM_180452.1 | NP_850783.1 | 16 - 17 | 2063 | 97 | ||
GenBank mRNA | AF360198.1 | - | 16 - 17 | 2063 | 97 | |
AY142616.1 | - | 16 - 17 | 1952 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |