Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125740.2 | NP_001119212.1 | 6 - 7 | 2161 | 74 | |
NM_121246.5 | NP_196769.1 | 6 - 7 | 2221 | 74 | ||
NM_180479.2 | NP_850810.1 | 5 - 6 | 2334 | 74 | ||
GenBank mRNA | AK318910.1 | - | 6 - 7 | 2095 | 74 | |
AY075640.1 | - | 6 - 7 | 2144 | 74 | ||
BT002236.1 | - | 4 - 5 | 1961 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |