Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121634.4 | NP_197133.1 | 6 - 7 | 943 | 146 | |
NM_180498.3 | NP_850829.1 | 6 - 7 | 930 | 146 | ||
GenBank mRNA | AK226906.1 | - | 6 - 7 | 869 | 146 | |
BT021989.1 | - | 6 - 7 | 832 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |