Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180596.2 | NP_850927.1 | 2 - 3 | 791 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |