Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198248.1 | NP_001185177.1 | 13 - 14 | 1765 | 93 | |
NM_001333364.1 | NP_001320175.1 | 13 - 14 | 1780 | 93 | ||
NM_103797.4 | NP_175333.3 | 13 - 14 | 1717 | 93 | ||
NM_180628.4 | NP_850959.1 | 13 - 14 | 1803 | 93 | ||
GenBank mRNA | AY082605.1 | - | 13 - 14 | 1610 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |